(S)-(+)-2-Butanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (S)-(+)-sec-Butyl alcohol, (S)-Butan-2-ol, (S)-(+)-sec-Butanol, (S)-(+)-Ethyl methyl carbinol, (S)-(+)-2-Hydroxy butane, (S)-(+)-Methyl ethyl carbinol

B0925-1ML B0925-5ML
TCB0925-1MLEA 49 USD
TCB0925-1ML TCB0925-5ML
(S)-(+)-2-Butanol ≥98.0% (by GC)
(S)-(+)-2-Butanol

CAS Number: 4221-99-2
MDL Number: MFCD00064281
Molecular Formula: C4H10O
Molecular Weight: 74.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 24
Specific Gravity (20/20): 0.81
Specific rotation [a]20/D: 13 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: (S)-(+)-sec-Butyl Alcohol

Formula: C₄H₁₀O
MW: 74.12 g/mol
Boiling Pt: 100 °C
Density: 0.81 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00064281
CAS Number: 4221-99-2
UN: 1120
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de5a55d613305eb6b10dc2f84ad71cb456b1a4bc&vid=de5a55d613305eb6b10dc2f84ad71cb456b1a4bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR