1-Bromo-3-fluorobenzene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
3-Bromofluorobenzene
B0882-250G
B0882-25G
TCB0882-250GEA
207
USD
TCB0882-250G
TCB0882-25G
1-Bromo-3-fluorobenzene ≥98.0% (by GC)
1-Bromo-3-fluorobenzene
CAS Number: 1073-06-9
MDL Number: MFCD00000326
Molecular Formula: C6H4BrF
Molecular Weight: 175.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Flash Point (°C): 39
Specific Gravity (20/20): 1.60
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 1-Bromo-3-fluorobenzene
Formula:
C₆H₄BrF MW: 175 g/mol Boiling Pt: 147 °C Density: 1.60 (20 °C) Flash Pt: 39 °C |
MDL Number:
MFCD00000326 CAS Number: 1073-06-9 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...