1,3-Benzenedisulfonyl chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 585-47-7
MDL Number: MFCD00041879
Molecular Formula: C6H4Cl2O4S2
Molecular Weight: 275.11
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 62
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1,3-Benzenedisulfonyl Dichloride
Formula:
C₆H₄Cl₂O₄S₂ Boiling Pt: 145 °C Melting Pt: 62 °C |
MDL Number:
MFCD00041879 CAS Number: 585-47-7 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...