α-Bromo-p-toluic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-Bromomethylbenzoic acid, 4-(Bromomethyl)benzoic acid
B0845-25G
B0845-5G
TCB0845-25GEA
67.13
USD
TCB0845-25G
TCB0845-5G
α-Bromo-p-toluic acid ≥97.0% (by GC, titration analysis)
α-Bromo-p-toluic acid
CAS Number: 6232-88-8
MDL Number: MFCD00002567
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 230
Other Product Names / Synonyms: alpha-Bromo-p-toluic Acid
Formula:
C₈H₇BrO₂ MW: 215.05 g/mol Melting Pt: 230 °C |
MDL Number:
MFCD00002567 CAS Number: 6232-88-8 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...