N,O-Bis(trimethylsilyl)trifluoroacetamide ≥95.0%

Supplier: TCI America

Synonyms: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA

B0830-25ML B0830-5ML B0830-100ML
TCB0830-25MLEA 38.48 USD
TCB0830-25ML TCB0830-5ML TCB0830-100ML
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥95.0%
N,O-Bis(trimethylsilyl)trifluoroacetamide

CAS Number: 25561-30-2
MDL Number: MFCD00008269
Molecular Formula: C8H18F3NOSi2
Molecular Weight: 257.40
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 24
Specific Gravity (20/20): 0.97
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis), Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: BSTFA

Formula: C₈H₁₈F₃NOSi₂
Boiling Pt: 142 °C
Density: 0.97 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00008269
CAS Number: 25561-30-2
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c99fa185b97f6241bc433a6ee06039fdd3d44d9b&vid=c99fa185b97f6241bc433a6ee06039fdd3d44d9b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR