3,3-Dimethylbutyric acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3,3-Dimethylbutanoic acid, tert-Butylacetic acid
B0826-25ML
B0826-500ML
TCB0826-25MLEA
49
USD
TCB0826-25ML
TCB0826-500ML
3,3-Dimethylbutyric acid ≥98.0% (by GC, titration analysis)
3,3-Dimethylbutyric acid
CAS Number: 1070-83-3
MDL Number: MFCD00002715
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 190
Melting point (°C): -11
Flash Point (°C): 89
Specific Gravity (20/20): 0.91
Other Product Names / Synonyms: 3,3-Dimethylbutyric Acid
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 190 °C Melting Pt: -11 °C Density: 0.91 (20 °C) Flash Pt: 89 °C |
MDL Number:
MFCD00002715 CAS Number: 1070-83-3 UN: 3265 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...