2-Butyne-1-ol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Butyn-1-ol, But-2-yn-1-ol, 1-Hydroxy-2-butyne

B0712-25ML B0712-5ML
TCB0712-25MLEA 111 USD
TCB0712-25ML TCB0712-5ML
2-Butyne-1-ol ≥98.0% (by GC)
2-Butyne-1-ol

CAS Number: 764-01-2
MDL Number: MFCD00002914
Molecular Formula: C4H6O
Molecular Weight: 70.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 55
Specific Gravity (20/20): 0.94
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Hydroxymethylmethylacetylene

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 145 °C
Density: 0.94 (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00002914
CAS Number: 764-01-2
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=160ea8a895cb1772d01dca25a9ee80d1cd0793fb&vid=160ea8a895cb1772d01dca25a9ee80d1cd0793fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR