3-Bromopyridine ≥98.0% (by GC) stabilized

Supplier: TCI America

B0651-25G B0651-100G
TCB0651-25GEA 42.72 USD
TCB0651-25G TCB0651-100G
3-Bromopyridine ≥98.0% (by GC) stabilized
3-Bromopyridine

(stabilized with Copper chip)
CAS Number: 626-55-1
MDL Number: MFCD00006373
Molecular Formula: C5H4BrN
Molecular Weight: 158.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Melting point (°C): -27
Flash Point (°C): 52
Specific Gravity (20/20): 1.62

Formula: C₅H₄BrN
Boiling Pt: 178 °C
Melting Pt: –27 °C
Density: 1.62 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00006373
CAS Number: 626-55-1
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f62ce5ecd359cd64e481af6449b0032b2b18ba5c&vid=f62ce5ecd359cd64e481af6449b0032b2b18ba5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR