ω-Bromo-4-phenylacetophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B0633-25G B0633-5G
TCB0633-25GEA 121 USD
TCB0633-25G TCB0633-5G
ω-Bromo-4-phenylacetophenone ≥98.0% (by GC, titration analysis)
ω-Bromo-4-phenylacetophenone

CAS Number: 135-73-9
MDL Number: MFCD00000202
Molecular Formula: C14H11BrO
Molecular Weight: 275.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 125


Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-Bromoacetylbiphenyl, 4-Phenylphenacyl Bromide

Formula: C₁₄H₁₁BrO
Melting Pt: 125 °C
MDL Number: MFCD00000202
CAS Number: 135-73-9
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e8f1a79fc9a2b557224a98b44d2e507dc9fc3b1&vid=7e8f1a79fc9a2b557224a98b44d2e507dc9fc3b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR