2-Bromophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B0630-25G B0630-500G B0630-100G
TCB0630-25GEA 43.07 USD
TCB0630-25G TCB0630-500G TCB0630-100G
2-Bromophenol ≥98.0% (by GC, titration analysis)
2-Bromophenol

CAS Number: 95-56-7
MDL Number: MFCD00002146
Molecular Formula: C6H5BrO
Molecular Weight: 173.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 194
Flash Point (°C): 42
Freezing point (°C): 3
Specific Gravity (20/20): 1.64

Formula: C₆H₅BrO
MW: 173.01 g/mol
Boiling Pt: 194 °C
Density: 1.64 (20 °C)
Flash Pt: 42 °C
MDL Number: MFCD00002146
CAS Number: 95-56-7
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4313603ad2496fd91933a5345fafa66bb64b9976&vid=0832ed3e89c6ac231457a897e9e9e91fd29e9da2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR