1-Bromo-4-nitrobenzene ≥99.0% (by GC)

Supplier: TCI America

Synonyms: 4-Bromonitrobenzene

B0624-250G B0624-25G
TCB0624-250GEA 196 USD
TCB0624-250G TCB0624-25G
1-Bromo-4-nitrobenzene ≥99.0% (by GC)
1-Bromo-4-nitrobenzene

CAS Number: 586-78-7
MDL Number: MFCD00007280
Molecular Formula: C6H4BrNO2
Molecular Weight: 202.01
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 127
Flash Point (°C): 110


Other Product Names / Synonyms: 4-Bromonitrobenzene

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 256 °C
Melting Pt: 127 °C
Flash Pt: 110 °C
MDL Number: MFCD00007280
CAS Number: 586-78-7
UN: 3459
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ebd1d07cef981b215dcdad467fb333d432be44f&vid=6ad92b0bf05f245ffa74e3b382fd6c31ec3e235a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR