2-Bromoethyl ethyl ether ≥95.0% (by GC)
Supplier: TCI America
CAS Number: 592-55-2
MDL Number: MFCD00000237
Molecular Formula: C4H9BrO
Molecular Weight: 153.02
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 21
Specific Gravity (20/20): 1.37
Storage Temperature: <0°C
Other Product Names / Synonyms: 2-Bromodiethyl Ether, 2-Ethoxyethyl Bromide
Formula:
C₄H₉BrO Boiling Pt: 126 °C Density: 1.37 (20 °C) Flash Pt: 21 °C |
MDL Number:
MFCD00000237 CAS Number: 592-55-2 UN: 2340 ADR: 3,II |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...