α-Bromo-4-chlorotoulene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
alpha-Bromo-4-chlorotoluene, 1-(Bromomethyl)-4-chlorobenzene, 4-Chlorobenzyl bromide
B0576-25G
B0576-5G
TCB0576-25GEA
91
USD
TCB0576-25G
TCB0576-5G
α-Bromo-4-chlorotoulene ≥98.0% (by GC)
α-Bromo-4-chlorotoulene
CAS Number: 622-95-7
MDL Number: MFCD00040714
Molecular Formula: C7H6BrCl
Molecular Weight: 205.48
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 110
Melting point (°C): 50
Storage Temperature: 0-10°C
Other Product Names / Synonyms: alpha-Bromo-4-chlorotoluene
Formula:
C₇H₆BrCl MW: 205.48 g/mol Boiling Pt: 110 °C Melting Pt: 50 °C |
MDL Number:
MFCD00040714 CAS Number: 622-95-7 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...