4-Bromobenzoyl chloride ≥98.0%

Supplier: TCI America

Synonyms: p-Bromobenzoyl chloride

B0558-250G B0558-25G
TCB0558-250GEA 348 USD
TCB0558-250G TCB0558-25G
4-Bromobenzoyl chloride ≥98.0%
4-Bromobenzoyl chloride

CAS Number: 586-75-4
MDL Number: MFCD00000683
Molecular Formula: C7H4BrClO
Molecular Weight: 219.46
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 247
Melting point (°C): 38
Flash Point (°C): 110

Formula: C₇H₄BrClO
MW: 219.46 g/mol
Boiling Pt: 247
Melting Pt: 36
Flash Pt: 110 °C
MDL Number: MFCD00000683
CAS Number: 586-75-4
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5f64575b6a4c7c698e902c9efb41dbc921b2f3f&vid=f5f64575b6a4c7c698e902c9efb41dbc921b2f3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR