3-Bromobenzoyl chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
m-Bromobenzoyl chloride, 3-Bromobenzoylchloride
B0557-25G
B0557-5G
TCB0557-25GEA
101
USD
TCB0557-25G
TCB0557-5G
3-Bromobenzoyl chloride ≥98.0% (by titrimetric analysis)
3-Bromobenzoyl chloride
CAS Number: 1711-09-7
MDL Number: MFCD00000669
Molecular Formula: C7H4BrClO
Molecular Weight: 219.46
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 240
Flash Point (°C): 107
Specific Gravity (20/20): 1.67
Formula:
C₇H₄BrClO MW: 219.46 g/mol Boiling Pt: 240 °C Density: 1.67 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00000669 CAS Number: 1711-09-7 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...