2-Bromoaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B0541-250G B0541-25G
TCB0541-250GEA 255 USD
TCB0541-250G TCB0541-25G
2-Bromoaniline ≥98.0% (by GC, titration analysis)
2-Bromoaniline

CAS Number: 615-36-1
MDL Number: MFCD00007632
Molecular Formula: C6H6BrN
Molecular Weight: 172.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 229
Flash Point (°C): 110
Freezing point (°C): 29

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 229 °C
Flash Pt: 110 °C
MDL Number: MFCD00007632
CAS Number: 615-36-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2501016c67dcd17f771dc5f346f4abce627d16cb&vid=47c6bdc21f5690f0ad625f5d6f106cb50f054429&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR