4,4'-Bipyridyl ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4,4'-Bipyridine, 4,4'-Dipyridyl

B0469-100G B0469-25G
TCB0469-100GEA 253 USD
TCB0469-100G TCB0469-25G
4,4'-Bipyridyl ≥98.0% (by GC, titration analysis)
4,4'-Bipyridyl

CAS Number: 553-26-4
MDL Number: MFCD00006416
Molecular Formula: C10H8N2
Molecular Weight: 156.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 161
Melting point (°C): 112
Lambda max.: 300 nm (CH3CN)


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 4,4'-Bipyridine, 4,4'-Dipyridyl

Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Boiling Pt: 161 °C
Melting Pt: 112 °C
MDL Number: MFCD00006416
CAS Number: 553-26-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbdb5d49cb29c7890675ce42653d3a6fe3e6a623&vid=cbdb5d49cb29c7890675ce42653d3a6fe3e6a623&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR