2,2'-Bipyridyl ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl
CAS Number: 366-18-7
MDL Number: MFCD00006212
Molecular Formula: C10H8N2
Molecular Weight: 156.19
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Boiling point (°C): 273
Melting point (°C): 72
Flash Point (°C): 121
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Ligands, Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Precise Polymerizatio
Other Product Names / Synonyms: 2,2'-Bipyridine, 2,2'-Dipyridyl
Formula:
C₁₀H₈N₂ MW: 156.19 g/mol Boiling Pt: 273 °C Melting Pt: 72 °C Flash Pt: 121 °C |
MDL Number:
MFCD00006212 CAS Number: 366-18-7 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >99.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...