Toluene-α-sulphonyl chloride ≥97.0%

Supplier: TCI America

Synonyms: Benzylsulphonyl chloride, α-Toluenesulphonyl chloride, Phenylmethylsulphonyl chloride, Phenylmethanesulphonyl chloride

B0441-25G B0441-100G
TCB0441-25GEA 78.39 USD
TCB0441-25G TCB0441-100G
Toluene-α-sulphonyl chloride ≥97.0%
Toluene-α-sulphonyl chloride

CAS Number: 1939-99-7
MDL Number: MFCD00007455
Molecular Formula: C7H7ClO2S
Molecular Weight: 190.64
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 92


Other Product Names / Synonyms: alpha-Toluenesulfonyl Chloride

Formula: C₇H₇ClO₂S
Boiling Pt: 120 °C
Melting Pt: 92 °C
MDL Number: MFCD00007455
CAS Number: 1939-99-7
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cf95d31f84f23d7ce93ac46fb8d11aa6b852dff&vid=9cf95d31f84f23d7ce93ac46fb8d11aa6b852dff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR