1,4-Butanesultone ≥99.0% (by GC)

Supplier: TCI America

Synonyms: 1,4-Butane sultone

B0136-250G B0136-25G
TCB0136-250GEA 268 USD
TCB0136-250G TCB0136-25G
1,4-Butanesultone ≥99.0% (by GC)
1,4-Butanesultone

CAS Number: 1633-83-6
MDL Number: MFCD00006584
Molecular Formula: C4H8O3S
Molecular Weight: 136.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 110
Freezing point (°C): 14
Specific Gravity (20/20): 1.34


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 1,2-Oxathiane 2,2-Dioxide

Formula: C₄H₈O₃S
MW: 136.17 g/mol
Boiling Pt: 146 °C
Density: 1.34 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00006584
CAS Number: 1633-83-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96b53df4aee4af05fcbe78e8e8e6e54eee8aa5cd&vid=96b53df4aee4af05fcbe78e8e8e6e54eee8aa5cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR