Tripropyl borate ≥99.0%

Supplier: TCI America

B0134-25ML B0134-500ML
TCB0134-25MLEA 46 USD
TCB0134-25ML TCB0134-500ML
Tripropyl borate ≥99.0%
Tripropyl borate

CAS Number: 688-71-1
MDL Number: MFCD00009366
Molecular Formula: C9H21BO3
Molecular Weight: 188.07
Purity/Analysis Method: >99.0% (T)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 32
Specific Gravity (20/20): 0.86


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Boric Acid Tripropyl Ester, Propyl Borate

Formula: C₉H₂₁BO₃
Boiling Pt: 180 °C
Density: 0.86 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00009366
CAS Number: 688-71-1
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edb70212835ccdb2635cbe2090241f07f6881cff&vid=edb70212835ccdb2635cbe2090241f07f6881cff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR