4,4'-TDE ≥98.0% (by GC)

Supplier: TCI America

B0132-5G B0132-1G
TCB0132-5GEA 130 USD
TCB0132-5G TCB0132-1G
4,4'-TDE ≥98.0% (by GC)
4,4'-TDE

CAS Number: 72-54-8
MDL Number: MFCD00000851
Molecular Formula: C14H10Cl4
Molecular Weight: 320.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 111


Product Categories: Analytical Science, Chlorinated Compounds (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: p,p'-DDD, 1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane

Formula: C₁₄H₁₀Cl₄
Melting Pt: 111 °C
MDL Number: MFCD00000851
CAS Number: 72-54-8
UN: 2761
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1ed63e1edc7a595f8d6704d363c3499c10e104&vid=8a1ed63e1edc7a595f8d6704d363c3499c10e104&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR