Undecafluorohexanoic acid ≥98.0% (by titrimetric analysis), high purity

Supplier: TCI America

A5722-1G A5722-5G
TCA5722-1GEA 101.85 USD
TCA5722-1G TCA5722-5G
Undecafluorohexanoic acid ≥98.0% (by titrimetric analysis), high purity
Undecafluorohexanoic acid

[Ion-Pair Reagent for LC-MS]
CAS Number: 307-24-4
MDL Number: MFCD00198040
Molecular Formula: C6HF11O2
Molecular Weight: 314.05
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 157
Melting point (°C): 14
Specific Gravity (20/20): 1.76


Product Categories: Analytical Science, LC/MS Ion-Pair Reagents for Basic Samples
Other Product Names / Synonyms: Perfluorohexanoic Acid High Grade, IPC-PFFA-6 HG

Formula: C₆HF₁₁O₂
Boiling Pt: 157 °C
Melting Pt: 14 °C
Density: 1.76 (20 °C)
MDL Number: MFCD00198040
CAS Number: 307-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46775735dac20378195be29377070336cb72b371&vid=46775735dac20378195be29377070336cb72b371&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR