Nɑ-(5-Fluoro-2,4-dinitrophenyl)-D-leucinamide ≥98.0% (by HPLC)
Supplier: TCI America
A5524-100MG
A5524-1G
TCA5524-100MGEA
145.56
USD
TCA5524-100MG
TCA5524-1G
Nɑ-(5-Fluoro-2,4-dinitrophenyl)-D-leucinamide ≥98.0% (by HPLC)
Nalpha-(5-Fluoro-2,4-dinitrophenyl)-D-leucinamide
[HPLC Labeling Reagent for e.e. Determination]
CAS Number: 178065-30-0
MDL Number: MFCD03844762
Molecular Formula: C12H15FN4O5
Molecular Weight: 314.27
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 170
Specific rotation [a]20/D: -4 deg (C=1, Acetone)
Product Categories: Analytical Science, e.e. Determination (HPLC Labeling Reagents), UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: Nalpha-(2,4-Dinitro-5-fluorophenyl)-D-leucinamide
Formula:
C₁₂H₁₅FN₄O₅ Melting Pt: 170 °C |
MDL Number:
MFCD03844762 CAS Number: 178065-30-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...