Sodium tetraphenylborate ≥99.5% (by gravimetric analysis)

Supplier: TCI America

Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt

A5130-10G A5130-1G
TCA5130-10GEA 100 USD
TCA5130-10G TCA5130-1G
Sodium tetraphenylborate ≥99.5% (by gravimetric analysis)
Sodium tetraphenylborate

[Precipitation reagent for K]
CAS Number: 143-66-8
MDL Number: MFCD00011494
Molecular Formula: C24H20BNa
Molecular Weight: 342.22
Purity/Analysis Method: >99.5% (W)
Form: Crystal


Product Categories: Analytical Science, Ion Association Reagents
Other Product Names / Synonyms: Tetraphenylboron Sodium

Formula: (C₆H₅)₄BNa
MW: 342.22 g/mol
MDL Number: MFCD00011494
CAS Number: 143-66-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77bb16f6af3a319132328866d2acfd29cf44ae74&vid=db0428e6a501fe5658276754bfe908dba9e54444&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.5% (W)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR