4-(2-Pyridylazo)resorcinol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-(2'-Pyridylazo)resorcinol

A5001-1G A5001-5G
TCA5001-1GEA 40 USD
TCA5001-1G TCA5001-5G
4-(2-Pyridylazo)resorcinol ≥98.0% (by HPLC, titration analysis)
4-(2-Pyridylazo)resorcinol

[Spectrophotometric reagent for transition metals]
CAS Number: 1141-59-9
MDL Number: MFCD00006256
Molecular Formula: C11H9N3O2
Molecular Weight: 215.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Analytical Science, Azo Compounds (Chelating Agents)
Other Product Names / Synonyms: 4-(2-Pyridylazo)resorcinol

Formula: C₁₁H₉N₃O₂
MW: 215.21
MDL Number: MFCD00044237
CAS Number: 1141-59-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fbf6ff8448eed375d38a86b45555bc7bb087ad8&vid=1258719c4f6567fb7939714b5087b2df460a5328&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR