Aminophylline ≥98.0%

Supplier: TCI America

Synonyms: 3,7-Dihydro-1,3-dimethylpurine-2,6-dione, complex with 1,2-ethanediamine (2:1), 1,3-Dimethyl-3,7-dihydro-1H-purine-2,6-dione hemiethane-1,2-diamine salt

A2805-100G A2805-25G
TCA2805-100GEA 86 USD
TCA2805-100G TCA2805-25G
Aminophylline ≥98.0%
Aminophylline

CAS Number: 317-34-0
MDL Number: MFCD00013221
Molecular Formula: C7H8N4O2
Molecular Weight: 420.43
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 272
Storage Temperature: <0°C

Formula: C₇H₈N₄O₂
MW: 420.43 g/mol
Melting Pt: 272 °C
MDL Number: MFCD00013221
CAS Number: 317-34-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e0b728dbb80ee32635908002d3547b8a5d8b086&vid=4e0b728dbb80ee32635908002d3547b8a5d8b086&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR