3-Amino-3-methyloxetane ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-Methyloxetan-3-amine, 3-Methyl-3-oxetanamine
A2792-1G
A2792-200MG
TCA2792-1GEA
391.3
USD
TCA2792-1G
TCA2792-200MG
3-Amino-3-methyloxetane ≥97.0% (by GC, titration analysis)
3-Amino-3-methyloxetane
CAS Number: 874473-14-0
MDL Number: MFCD11111713
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 97
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 3-Methyl-3-oxetanamine
Formula:
C₄H₉NO MW: 87.12 g/mol Density: 0.98 (20 °C) Flash Pt: 97 °C |
MDL Number:
MFCD11111713 CAS Number: 874473-14-0 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...