2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2773-1G A2773-5G
TCA2773-1GEA 40 USD
TCA2773-1G TCA2773-5G
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide ≥98.0% (by GC, titration analysis)
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide

CAS Number: 302964-24-5
MDL Number: MFCD10000630
Molecular Formula: C11H10ClN3OS
Molecular Weight: 267.73
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 210

Formula: C₁₁H₁₀ClN₃OS
MW: 267.73 g/mol
Melting Pt: 210 °C
MDL Number: MFCD10000630
CAS Number: 302964-24-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fa42a752cd2699ba84657be50289f88e54cd27a&vid=8fa42a752cd2699ba84657be50289f88e54cd27a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR