(Acetylacetonato)(norbornadiene)rhodium(I)

Supplier: TCI America

Synonyms: (Acetylacetonato)[bicyclo[2.2.1]hepta-2,5-diene]rhodium(I), (Norbornadiene)rhodium(I) 2,4-pentanedionate

A2761-1G A2761-200MG
TCA2761-1GEA 718 USD
TCA2761-1G TCA2761-200MG
(Acetylacetonato)(norbornadiene)rhodium(I)
(Acetylacetonato)(norbornadiene)rhodium(I)

CAS Number: 32354-50-0
MDL Number: MFCD00075051
Molecular Formula: C12H15O2Rh
Molecular Weight: 294.16
Form: Crystal
Color: Pale Yellow
Melting point (°C): 175


Other Product Names / Synonyms: (Acetylacetonato)[bicyclo[2.2.1]hepta-2,5-diene]rhodium(I)

Formula: C₁₂H₁₅O₂Rh
MW: 294.16 g/mol
Melting Pt: 175 °C
MDL Number: MFCD00075051
CAS Number: 32354-50-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9f2ef24e68a4a8db631f96582bc448ccd246cfb&vid=f9f2ef24e68a4a8db631f96582bc448ccd246cfb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR