13-Azido-2,5,8,11-tetraoxatridecane ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: mPEG4-Azide, O-(2-Azidoethyl)-O'-methyltriethylene Glycol, mPEG4-N3, Methyl-PEG4-Azide

A2728-100MG A2728-25MG
TCA2728-100MGEA 236 USD
TCA2728-100MG TCA2728-25MG
13-Azido-2,5,8,11-tetraoxatridecane ≥98.0% (by HPLC)
13-Azido-2,5,8,11-tetraoxatridecane

CAS Number: 606130-90-9
MDL Number: MFCD13184960
Molecular Formula: C9H19N3O4
Molecular Weight: 233.27
Purity/Analysis Method: >98.0% (HPLC)
Form: Clear Liquid
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Azide Compounds (Click Chemistry), Bioscience, PEGylation Reagents & PEG Spacers, PEGylation Reagents
Other Product Names / Synonyms: O-(2-Azidoethyl)-O'-methyltriethylene Glycol, mPEG4-Azide, mPEG4-N3, Methyl-PEG4-Azide

Formula: C₉H₁₉N₃O₄
MW: 233.27 g/mol
MDL Number: MFCD13184960
CAS Number: 606130-90-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d34d21b5de375e978e98022dc16c24f41437b2cc&vid=4c7ab3007240cbbc3ed729a52f31ac1fb0f939a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR