5'-Amino-2'-methylacetanilide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Acetamido-4-methylaniline, N-(5-Amino-2-methylphenyl)acetamide, N3-Acetyl-4-methyl-1,3-phenylenediamine

A2724-1G A2724-5G
TCA2724-1GEA 102 USD
TCA2724-1G TCA2724-5G
5'-Amino-2'-methylacetanilide ≥98.0% (by HPLC, titration analysis)
5'-Amino-2'-methylacetanilide

CAS Number: 5434-30-0
MDL Number: MFCD00047859
Molecular Formula: C9H12N2O
Molecular Weight: 164.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 144


Other Product Names / Synonyms: N3-Acetyl-4-methyl-1,3-phenylenediamine, 3-Acetamido-4-methylaniline

Formula: C₉H₁₂N₂O
MW: 164.21 g/mol
Melting Pt: 144 °C
MDL Number: MFCD00047859
CAS Number: 5434-30-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1948dacad50f42f91fe26033853525f3904cc459&vid=1948dacad50f42f91fe26033853525f3904cc459&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR