2-Allylisoindoline-1,3-dione ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
N-Allylphthalimide
A2696-1G
A2696-200MG
TCA2696-1GEA
188
USD
TCA2696-1G
TCA2696-200MG
2-Allylisoindoline-1,3-dione ≥98.0% (by GC)
2-Allylisoindoline-1,3-dione
CAS Number: 5428-09-1
MDL Number: MFCD00158662
Molecular Formula: C11H9NO2
Molecular Weight: 187.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 295
Melting point (°C): 70
Formula:
C₁₁H₉NO₂ MW: 187.19 g/mol Boiling Pt: 295 °C Melting Pt: 70 °C |
MDL Number:
MFCD00158662 CAS Number: 5428-09-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...