2-Amino-4-(trifluoromethyl)benzonitrile ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

A2648-1G A2648-5G
TCA2648-1GEA 51 USD
TCA2648-1G TCA2648-5G
2-Amino-4-(trifluoromethyl)benzonitrile ≥98.0% (by GC, total nitrogen)
2-Amino-4-(trifluoromethyl)benzonitrile

CAS Number: 1483-54-1
MDL Number: MFCD04974124
Molecular Formula: C8H5F3N2
Molecular Weight: 186.14
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Boiling point (°C): 127
Melting point (°C): 90


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Amino-4-cyanobenzotrifluoride, 2-Cyano-5-(trifluoromethyl)aniline, 4-(Trifluoromethyl)anthranilonitrile

Formula: C₈H₅F₃N₂
Boiling Pt: 127 °C
Melting Pt: 90 °C
MDL Number: MFCD04974124
CAS Number: 1483-54-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=062dcfd39932c8705555644bdac0c01be0bb619e&vid=062dcfd39932c8705555644bdac0c01be0bb619e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR