4-Amino-3-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 327-74-2
MDL Number: MFCD00275473
Molecular Formula: C8H5F3N2
Molecular Weight: 186.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 62
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Amino-5-cyanobenzotrifluoride, 4-Cyano-2-(trifluoromethyl)aniline
Formula:
C₈H₅F₃N₂ Boiling Pt: 100 °C Melting Pt: 62 °C |
MDL Number:
MFCD00275473 CAS Number: 327-74-2 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...