Acrylamide ∼50% in water

Supplier: TCI America

Synonyms: Prop-2-enamide

A2625-500G A2625-100G
TCA2625-500GEA 53.31 USD
TCA2625-500G TCA2625-100G
Acrylamide ∼50% in water
Acrylamide (monomer)

CAS Number: 79-06-1
MDL Number: MFCD00008032
Molecular Formula: C3H5NO
Molecular Weight: 71.08
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 106
Storage Temperature: 0-10°C

Formula: CH₂=CHCONH₂
MW: 71.08 g/mol
Boiling Pt: 106 °C
MDL Number: MFCD00008032
CAS Number: 79-06-1
UN: 2074
ADR: 6.1,III
https://pr.vwr.com/stibo/web/std.lang.all/16/22/28931622.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR