4-(2-Aminoethyl)cyclohexylamine (cis- and trans- mixture) ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-(4-Aminocyclohexyl)ethylamine, 1-Amino-4-(2-aminoethyl)cyclohexane
A2609-1G
A2609-5G
TCA2609-1GEA
233
USD
TCA2609-1G
TCA2609-5G
4-(2-Aminoethyl)cyclohexylamine (cis- and trans- mixture) ≥97.0% (by titrimetric analysis)
4-(2-Aminoethyl)cyclohexylamine (cis- and trans- mixture)
CAS Number: 202256-86-8
Molecular Formula: C8H18N2
Molecular Weight: 142.25
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 129
Other Product Names / Synonyms: 2-(4-Aminocyclohexyl)ethylamine, 1-Amino-4-(2-aminoethyl)cyclohexane
Formula:
C₈H₁₈N₂ MW: 142.25 g/mol Boiling Pt: 129 °C |
CAS Number:
202256-86-8 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...