2-Allyl-5,5-dimethyl-1,3,2-dioxaborinane ≥98.0% (by GC, titration analysis) stabilized

Supplier: TCI America

A2574-1G A2574-5G
TCA2574-1GEA 138.1 USD
TCA2574-1G TCA2574-5G
2-Allyl-5,5-dimethyl-1,3,2-dioxaborinane ≥98.0% (by GC, titration analysis) stabilized
2-Allyl-5,5-dimethyl-1,3,2-dioxaborinane

(stabilized with Phenothiazine)
CAS Number: 911482-75-2
Molecular Formula: C8H15BO2
Molecular Weight: 154.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.93
Storage Temperature: 0-10°C

Formula: C₈H₁₅BO₂
MW: 154.02 g/mol
Density: 0.93 (20 °C)
CAS Number: 911482-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1c67d74dfc8812b533cece9d0034aa50563902f&vid=d1c67d74dfc8812b533cece9d0034aa50563902f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR