Amprolium hydrochloride ≥98.0% (by HPLC, titration analysis) for biochemical research

Supplier: TCI America

Synonyms: Amprolium HCl, 1-([4-Amino-2-propyl-5-pyrimidinyl]methyl)-2-methylpyridinium chloride, Amprol, 1-((4-Amino-2-propylpyrimidin-5-yl)methyl)-2-methylpyridin-1-ium chloride hydrochloride, Amprolium hydrochloride, 1-[(4-Amino-2-propyl-5-pyrimidinyl)methyl]-2-methylpyridinium Chloride Hydrochloride, Mepyrium, Thiacoccid

A2572-25G A2572-5G
TCA2572-25GEA 71 USD
TCA2572-25G TCA2572-5G
Amprolium hydrochloride ≥98.0% (by HPLC, titration analysis) for biochemical research
Amprolium hydrochloride

[for Biochemical Research]
CAS Number: 137-88-2
MDL Number: MFCD00078831
Molecular Formula: C14H19ClN4
Molecular Weight: 315.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 241


Product Categories: Bioscience, Preservatives for Biochemical Research, Anthelmintic Ingredients
Other Product Names / Synonyms: 1-[(4-Amino-2-propyl-5-pyrimidinyl)methyl]-2-methylpyridinium Chloride Hydrochloride

Formula: C₁₄H₁₉ClN₄
MW: 315.24 g/mol
Melting Pt: 241 °C
MDL Number: MFCD00078831
CAS Number: 137-88-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb646d38d60c298c5102716f7fe63e58cb108143&vid=a06046c7b1d8dbd71385d80266a8325c675c5587&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR