2-(Aminomethyl)-1-ethylpyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2556-25G A2556-5G
TCA2556-25GEA 130.8 USD
TCA2556-25G TCA2556-5G
2-(Aminomethyl)-1-ethylpyrrolidine ≥98.0% (by GC, titration analysis)
2-(Aminomethyl)-1-ethylpyrrolidine

CAS Number: 26116-12-1
MDL Number: MFCD00003178
Molecular Formula: C7H16N2
Molecular Weight: 128.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 56
Specific Gravity (20/20): 0.90

Formula: C₇H₁₆N₂
MW: 128.22 g/mol
Boiling Pt: 60 °C
Density: 0.90 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00003178
CAS Number: 26116-12-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1290f5d51ea86e17d169fab0a3e23429adc9a67&vid=f1290f5d51ea86e17d169fab0a3e23429adc9a67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR