4-(Aminomethyl)tetrahydropyran ≥98.0%
Supplier: TCI America
Synonyms:
4-(Aminomethyl)tetrahydro-2H-pyran, 4-Aminomethyltetrahydropyran, Tetrahydro-2H-pyran-4-ylmethanamine, (Tetrahydro-2H-pyran-4-yl)methanamine
A2550-1G
A2550-5G
TCA2550-1GEA
75
USD
TCA2550-1G
TCA2550-5G
4-(Aminomethyl)tetrahydropyran ≥98.0%
4-(Aminomethyl)tetrahydropyran
CAS Number: 130290-79-8
MDL Number: MFCD02179435
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 76
Specific Gravity (20/20): 0.97
Formula:
C₆H₁₃NO Boiling Pt: 76 °C Density: 0.97 (20 °C) |
MDL Number:
MFCD02179435 CAS Number: 130290-79-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...