Adapalene ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic Acid, 6-(3-(Adamantan-1-yl)-4-methoxyphenyl)-2-naphthoic acid

A2549-1G A2549-200MG
TCA2549-1GEA 365 USD
TCA2549-1G TCA2549-200MG
Adapalene ≥98.0% (by HPLC, titration analysis)
Adapalene

CAS Number: 106685-40-9
MDL Number: MFCD03106112
Molecular Formula: C28H28O3
Molecular Weight: 412.53
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 324
Storage Temperature: 0-10°C


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic Acid

Formula: C₂₈H₂₈O₃
MW: 412.53 g/mol
Melting Pt: 324 °C
MDL Number: MFCD03106112
CAS Number: 106685-40-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=487e0c470b87df3d672d31e5706398109f13f41d&vid=487e0c470b87df3d672d31e5706398109f13f41d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR