Atovaquone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthalenedione, 2-(trans-4-(4-Chlorophenyl)cyclohexyl)-3-hydroxynaphthalene-1,4-dione

A2545-1G A2545-200MG
TCA2545-1GEA 427.19 USD
TCA2545-1G TCA2545-200MG
Atovaquone ≥98.0% (by HPLC, titration analysis)
Atovaquone

CAS Number: 95233-18-4
MDL Number: MFCD00889188
Molecular Formula: C22H19ClO3
Molecular Weight: 366.84
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 223
Lambda max.: 494 nm (aq. Buffer)Storage Temperature: <0°C


Product Categories: Bioscience, Antifungal Ingredients, Antiprotozoal Ingredients
Other Product Names / Synonyms: trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthalenedione

Formula: C₂₂H₁₉ClO₃
MW: 366.84 g/mol
Melting Pt: 223 °C
MDL Number: MFCD00889188
CAS Number: 95233-18-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a806214042f8467b2ef1747046d6f0dd983c7378&vid=a806214042f8467b2ef1747046d6f0dd983c7378&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR