3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Amino-3-(3,4-dimethoxyphenyl)propionic acid

A2536-1G A2536-5G
TCA2536-1GEA 33.5 USD
TCA2536-1G TCA2536-5G
3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid

CAS Number: 34841-09-3
MDL Number: MFCD00735172
Molecular Formula: C11H15NO4
Molecular Weight: 225.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 228

Formula: C₁₁H₁₅NO₄
MW: 225.244 g/mol
Melting Pt: 228 °C
MDL Number: MFCD00735172
CAS Number: 34841-09-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5af16c97796ac501bdfcf646bdbf7f00e7b45d78&vid=5af16c97796ac501bdfcf646bdbf7f00e7b45d78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR