3-Amino-3-phenyl-1-propanol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

A2534-1G A2534-5G
TCA2534-1GEA 24 USD
TCA2534-1G TCA2534-5G
3-Amino-3-phenyl-1-propanol ≥97.0% (by GC, titration analysis)
3-Amino-3-phenyl-1-propanol

CAS Number: 14593-04-5
MDL Number: MFCD00801124
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 134
Melting point (°C): 75

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 134 °C
Melting Pt: 75 °C
MDL Number: MFCD00801124
CAS Number: 14593-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7479b384d5872091cf606fee9ae2207b777a99c0&vid=7479b384d5872091cf606fee9ae2207b777a99c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR