N-Allyl-N-tert-butylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-tert-Butylallylamine

A2514-100G A2514-25G
TCA2514-100GEA 772 USD
TCA2514-100G TCA2514-25G
N-Allyl-N-tert-butylamine ≥98.0% (by GC, titration analysis)
N-Allyl-N-tert-butylamine

CAS Number: 16486-68-3
Molecular Formula: C7H15N
Molecular Weight: 113.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 112
Specific Gravity (20/20): 0.76


Other Product Names / Synonyms: N-tert-Butylallylamine

Formula: C₇H₁₅N
MW: 113.20 g/mol
Boiling Pt: 112 °C
Density: 0.76 (20 °C)
CAS Number: 16486-68-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4b248a9373c31b8386a92ea478ca8ea0408bc0&vid=3a4b248a9373c31b8386a92ea478ca8ea0408bc0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR