Aripiprazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A2496-1G
A2496-5G
TCA2496-1GEA
74.25
USD
TCA2496-1G
TCA2496-5G
Aripiprazole ≥98.0% (by HPLC, titration analysis)
Aripiprazole
CAS Number: 129722-12-9
MDL Number: MFCD00892072
Molecular Formula: C23H27Cl2N3O2
Molecular Weight: 448.39
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 149
Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone
Formula:
C₂₃H₂₇Cl₂N₃O₂ Melting Pt: 149 °C |
MDL Number:
MFCD00892072 CAS Number: 129722-12-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...