1-Amino-1-cyclopropanecarbonitrile hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
A2493-1G
A2493-5G
TCA2493-1GEA
37
USD
TCA2493-1G
TCA2493-5G
1-Amino-1-cyclopropanecarbonitrile hydrochloride ≥98.0% (by total nitrogen and titration analysis)
1-Amino-1-cyclopropanecarbonitrile hydrochloride
CAS Number: 127946-77-4
MDL Number: MFCD04114063
Molecular Formula: C4H6N2
Molecular Weight: 118.56
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 220
Other Product Names / Synonyms: 1-Aminocyclopropyl Cyanide Hydrochloride, 1-Amino-1-cyanocyclopropane Hydrochloride, 1-Cyanocyclopropylamine Hydrochloride
Formula:
C₄H₇ClN₂ MW: 118.56 g/mol Melting Pt: 220 °C |
MDL Number:
MFCD04114063 CAS Number: 127946-77-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (N,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...