Acarbose ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Prandase, Acarbose Hydrate, 4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose, Precose, (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4S,5S,6R)-3,4-Dihydroxy-6-methyl-5-(((1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl)amino)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal, Glucobay

A2485-1G A2485-5G
TCA2485-1GEA 78 USD
TCA2485-1G TCA2485-5G
Acarbose ≥98.0% (by HPLC, total nitrogen)
Acarbose

CAS Number: 56180-94-0
MDL Number: MFCD00869592
Molecular Formula: C25H43NO18
Molecular Weight: 645.61
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Specific rotation [a]20/D: 176 deg (C=1, H2O)

Formula: C₂₅H₄₃NO₁₈
MW: 645.61 g/mol
MDL Number: MFCD00869592
CAS Number: 56180-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c584a83ff0bfcae70c3414fc73b0ff7af46d4dc4&vid=c584a83ff0bfcae70c3414fc73b0ff7af46d4dc4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR