2-Amino-4-chlorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America

A2483-25G A2483-5G
TCA2483-25GEA 187.3 USD
TCA2483-25G TCA2483-5G
2-Amino-4-chlorobenzonitrile ≥98.0% (by GC)
2-Amino-4-chlorobenzonitrile

CAS Number: 38487-86-4
MDL Number: MFCD00035927
Molecular Formula: C7H5ClN2
Molecular Weight: 152.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 163


Other Product Names / Synonyms: 5-Chloro-2-cyanoaniline, 4-Chloroanthranilonitrile

Formula: C₇H₅ClN₂
Melting Pt: 163 °C
MDL Number: MFCD00035927
CAS Number: 38487-86-4
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=891126b308eb4ea936b209a1f8f20126d8c9d61b&vid=891126b308eb4ea936b209a1f8f20126d8c9d61b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR